Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009921.2 | NP_001009921.1 | 15 | 1367 | ||
NM_015303.3 | NP_056118.2 | 14 | 1361 | |||
XM_005247251.3 | XP_005247308.1 | 15 | 1635 | |||
XM_005247253.4 | XP_005247310.1 | 15 | 1373 | |||
XM_006713556.3 | XP_006713619.1 | 7 | 655 | |||
XM_011512599.2 | XP_011510901.1 | 2 | 117 | |||
XM_011512600.2 | XP_011510902.1 | 7 | 569 | |||
XM_011512601.2 | XP_011510903.1 | 7 | 655 | |||
XM_017006040.1 | XP_016861529.1 | 13 | 1055 | |||
XM_017006041.1 | XP_016861530.1 | 15 | 1373 | |||
GenBank mRNA | AB018347.1 | - | 7 | 546 | ||
AK056661.1 | - | 15 | 1339 | |||
AK292423.1 | - | 14 | 1333 | |||
AL833838.1 | - | 12 | 900 | |||
BC140768.1 | - | 15 | 1303 | 9021636 | ||
BX647915.1 | - | 14 | 1322 |