Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009921.2 | NP_001009921.1 | 12 | 1055 | ||
NM_015303.3 | NP_056118.2 | 11 | 1049 | |||
XM_005247251.3 | XP_005247308.1 | 12 | 1323 | |||
XM_005247253.4 | XP_005247310.1 | 12 | 1061 | |||
XM_006713556.3 | XP_006713619.1 | 4 | 343 | |||
XM_011512600.2 | XP_011510902.1 | 4 | 257 | |||
XM_011512601.2 | XP_011510903.1 | 4 | 343 | |||
XM_017006040.1 | XP_016861529.1 | 10 | 743 | |||
XM_017006041.1 | XP_016861530.1 | 12 | 1061 | |||
GenBank mRNA | AB018347.1 | - | 4 | 234 | ||
AK056661.1 | - | 12 | 1027 | |||
AK292423.1 | - | 11 | 1021 | |||
AL833838.1 | - | 9 | 588 | |||
BC140768.1 | - | 12 | 991 | 9021636 | ||
BX647915.1 | - | 11 | 1011 |