Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsku | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009965.2 | NP_001009965.1 | 2 | 2207 | ||
XM_006229759.2 | XP_006229821.1 | 2 | 2219 | |||
XM_006229760.2 | XP_006229822.1 | 2 | 3100 | |||
XM_006229761.2 | XP_006229823.1 | 2 | 2402 | |||
GenBank mRNA | AY533242.1 | - | 2 | 2128 | ||
BC100644.1 | - | 2 | 2207 | 7930712 |