Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017908.3 | NP_060378.1 | 7 | 1919 | ||
XM_005259052.4 | XP_005259109.1 | 7 | 1920 | |||
XM_005259053.4 | XP_005259110.1 | 7 | 1803 | |||
XM_017026949.1 | XP_016882438.1 | 6 | 2005 | |||
GenBank mRNA | AK000633.1 | - | 7 | 1658 | ||
AK026153.1 | - | 6 | 1835 | |||
AL122108.1 | - | 3 | 901 | |||
AY279351.1 | - | 6 | 1574 | |||
AY280800.1 | - | 7 | 1698 | |||
BC017206.2 | - | 8 | 1556 | 3636172 |