Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010961.1 | NP_001010961.1 | 3 | 615 | ||
GenBank mRNA | BC089059.1 | - | 3 | 615 | 7320241 | |
FQ228748.1 | - | 3 | 273 |