Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012015.1 | NP_001012015.1 | 2 | 721 | ||
XM_006248595.2 | XP_006248657.1 | 6 | 1110 | |||
XM_008768886.1 | XP_008767108.1 | 7 | 1410 | |||
XM_008768887.1 | XP_008767109.1 | 3 | 718 | |||
GenBank mRNA | BC083787.1 | - | 2 | 721 | 7097568 |