Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dirc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012017.1 | NP_001012017.1 | 4 | 1078 | ||
GenBank mRNA | BC081960.1 | - | 4 | 1078 | 7124589 |