Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URGCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077663.2 | NP_001071131.1 | 6 | 1606 | ||
NM_001077664.2 | NP_001071132.1 | 6 | 1378 | |||
NM_001290075.1 | NP_001277004.1 | 6 | 1601 | |||
NM_001290076.1 | NP_001277005.1 | 7 | 1500 | |||
NM_017920.4 | NP_060390.3 | 5 | 1579 | |||
GenBank mRNA | AB040940.1 | - | 7 | 1903 | ||
AK026185.1 | - | 1 | 783 | |||
AK302459.1 | - | 7 | 1500 | |||
AK314339.1 | - | 5 | 1125 | |||
AL832134.1 | - | 4 | 3219 | |||
AY078404.1 | - | 5 | 1098 | |||
BC018426.1 | - | 5 | 1100 | 4213413 | ||
BX640797.1 | - | 6 | 1141 | |||
BX648505.1 | - | 6 | 1146 | |||
CR749398.1 | - | 6 | 1158 |