Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPLOC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017921.3 | NP_060391.2 | 14 | 1605 | ||
NR_130139.1 | 3 | 177 | ||||
XM_011524979.1 | XP_011523281.1 | 14 | 1600 | |||
XM_011524980.1 | XP_011523282.1 | 14 | 1615 | |||
XM_011524981.1 | XP_011523283.1 | 14 | 1615 | |||
XM_011524982.2 | XP_011523284.1 | 14 | 1615 | |||
XM_017024830.1 | XP_016880319.1 | 10 | 1001 | |||
GenBank mRNA | AB040932.1 | - | 14 | 1512 | ||
AK000664.1 | - | 14 | 1514 | |||
AK023046.1 | - | 12 | 1241 | |||
AK023272.1 | - | 11 | 1184 | |||
AK294473.1 | - | 13 | 1482 | |||
AL834300.1 | - | 11 | 1117 | |||
BC025930.1 | - | 14 | 1546 | 5017718 | ||
BU542911.1 | - | 3 | 145 | |||
HY179409.1 | - | 3 | 178 |