Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nuf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012028.1 | NP_001012028.1 | 2 | 214 | ||
GenBank mRNA | BC078993.1 | - | 2 | 214 | 7112621 |