Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012035.1 | NP_001012035.1 | 9 | 1612 | ||
XM_006250737.2 | XP_006250799.1 | 9 | 1740 | |||
XM_006250738.2 | XP_006250800.1 | 10 | 1849 | |||
XM_006250739.2 | XP_006250801.1 | 9 | 2240 | |||
XM_006250740.2 | XP_006250802.1 | 10 | 2301 | |||
XM_006250741.2 | XP_006250803.1 | 9 | 2192 | |||
XM_008770028.1 | XP_008768250.1 | 10 | 2349 | |||
XM_008770029.1 | XP_008768251.1 | 9 | 2196 | |||
GenBank mRNA | AY383681.1 | - | 9 | 1305 | ||
BC083590.1 | - | 9 | 1612 | 7132550 |