Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efemp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012039.1 | NP_001012039.1 | 5 | 336 | ||
XM_008770456.1 | XP_008768678.1 | 5 | 436 | |||
XM_008770457.1 | XP_008768679.1 | 4 | 356 | |||
GenBank mRNA | BC079455.1 | - | 5 | 336 | 7121509 | |
D89730.1 | - | 3 | 232 |