Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvrl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012064.1 | NP_001012064.1 | 8, 9 | 1540 | ||
GenBank mRNA | BC090075.1 | - | 8, 9 | 1540 | 7189358 |