Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012069.2 | NP_001012069.2 | 8 | 1164 | ||
GenBank mRNA | BC083920.1 | - | 8 | 1150 | 7135090 |