Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012083.1 | NP_001012083.1 | 2, 3 | 338 | ||
XM_006233234.2 | XP_006233296.1 | 2, 3 | 1933 | |||
XM_006233235.2 | XP_006233297.1 | 2, 3 | 1934 | |||
GenBank mRNA | BC082033.1 | - | 2, 3 | 338 | 7130739 |