Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cytip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012086.1 | NP_001012086.1 | 8 | 1574 | ||
XM_006234202.2 | XP_006234264.1 | 9 | 1548 | |||
GenBank mRNA | BC088207.1 | - | 8 | 1574 | 7381695 |