Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012091.1 | NP_001012091.1 | 1 | 1005 | ||
GenBank mRNA | BC089055.1 | - | 1 | 1005 | 7308908 |