Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xkr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012092.1 | NP_001012092.1 | 3 | 1774 | ||
XM_008762334.1 | XP_008760556.1 | 4 | 3498 | |||
GenBank mRNA | AY534261.1 | - | 3 | 1774 |