Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xkr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012099.1 | NP_001012099.1 | 3 | 1639 | ||
GenBank mRNA | AY534262.1 | - | 3 | 1639 | ||
BC105851.1 | - | 3 | 766 | 7191341 |