Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012100.1 | NP_001012100.1 | 1 | 668 | ||
GenBank mRNA | BC085796.1 | - | 1 | 668 | 7109760 |