Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNHIT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170670.1 | NP_001164141.1 | 6 | 984 | ||
NM_017953.3 | NP_060423.3 | 5 | 1101 | |||
XM_011541614.2 | XP_011539916.1 | 5 | 1115 | |||
GenBank mRNA | AF308296.1 | - | 5 | 748 | ||
AK000736.1 | - | 5 | 473 | |||
AK000767.1 | - | 5 | 1010 | |||
AK298149.1 | - | 6 | 984 | |||
AK314566.1 | - | 5 | 1029 | |||
AL442074.1 | - | 5 | 1086 | |||
BC026236.1 | - | 5 | 1088 | 4664103 | ||
BC110898.1 | - | 5 | 1019 | 6012059 |