Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012128.2 | NP_001012128.1 | 4 | 1596 | ||
GenBank mRNA | BC078921.1 | - | 3 | 1395 | ||
BC088185.1 | - | 3 | 1432 | 7367875 | ||
FQ220569.1 | - | 1 | 881 |