Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012142.1 | NP_001012142.1 | 3 | 353 | ||
XM_006247342.2 | XP_006247404.1 | 3 | 582 | |||
XM_008768079.1 | XP_008766301.1 | 3 | 1866 | |||
GenBank mRNA | BC078939.1 | - | 3 | 353 | 7108372 |