Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012145.1 | NP_001012145.1 | 10 | 878 | ||
GenBank mRNA | BC078948.1 | - | 10 | 878 | 7109741 |