Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012150.1 | NP_001012150.1 | 9, 10 | 1383 | ||
XM_006250393.2 | XP_006250455.1 | 10, 11 | 2451 | |||
XM_006250400.2 | XP_006250462.1 | 10, 11 | 1728 | |||
XM_006250401.2 | XP_006250463.1 | 10, 11 | 1728 | |||
XM_008769859.1 | XP_008768081.1 | 12, 13 | 2416 | |||
XM_008769860.1 | XP_008768082.1 | 10, 11 | 2977 | |||
XM_008769861.1 | XP_008768083.1 | 10, 11 | 2347 | |||
GenBank mRNA | BC083927.1 | - | 9, 10 | 1383 | 7135538 |