Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012155.1 | NP_001012155.1 | 1 | 636 | ||
XM_006251990.2 | XP_006252052.1 | 2 | 476 | |||
XM_006251991.2 | XP_006252053.1 | 2 | 492 | |||
XM_008770688.1 | XP_008768910.1 | 1 | 693 | |||
GenBank mRNA | BC081891.1 | - | 1 | 636 | 7109579 |