Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012275.2 | NP_001012275.1 | 8, 9 | 953 | ||
XM_006243784.2 | XP_006243846.1 | 8, 9 | 1421 | |||
XM_006243785.2 | XP_006243847.1 | 8, 9 | 1418 | |||
XM_006243787.2 | XP_006243849.1 | 7, 8 | 642 | |||
XM_008766537.1 | XP_008764759.1 | 8, 9 | 1106 | |||
XM_008766538.1 | XP_008764760.1 | 8, 9 | 1109 | |||
GenBank mRNA | CV126331.1 | - | 4, 5 | 457 |