Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012345.1 | NP_001012345.1 | 8 | 1309 | ||
XM_006229733.2 | XP_006229795.1 | 7 | 1349 | |||
XM_008759689.1 | XP_008757911.1 | 8 | 1060 | |||
GenBank mRNA | BC089846.1 | - | 8 | 1309 | 7372077 |