Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il21r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012469.1 | NP_001012487.1 | 5 | 691 | ||
XM_006230261.2 | XP_006230323.1 | 4 | 728 | |||
XM_006230262.2 | XP_006230324.1 | 5 | 704 | |||
GenBank mRNA | BC089857.1 | - | 5 | 691 | 7381492 | |
FQ226709.1 | - | 4 | 562 |