Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012631.1 | NP_001012649.1 | 8 | 1112 | ||
NM_001012632.1 | NP_001012650.1 | 7 | 911 | |||
NM_001012633.1 | NP_001012651.1 | 8 | 874 | |||
NM_001012634.1 | NP_001012652.1 | 6 | 852 | |||
NM_001012635.1 | NP_001012653.1 | 6 | 847 | |||
NM_001012636.1 | NP_001012654.1 | 5 | 835 | |||
NM_001012718.1 | NP_001012736.1 | 7 | 1048 | |||
NM_001308078.1 | NP_001295007.1 | 6 | 1046 | |||
NM_004221.4 | NP_004212.4 | 7 | 1045 | |||
GenBank mRNA | AK296143.1 | - | 6 | 857 | ||
W57940.1 | - | 1 | 377 |