Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRAMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012642.2 | NP_001012660.1 | 11 | 1046 | ||
XM_011521327.2 | XP_011519629.1 | 11 | 1155 | |||
XM_011521328.2 | XP_011519630.1 | 11 | 1506 | |||
XM_011521329.2 | XP_011519631.1 | 11 | 1023 | |||
XM_011521330.2 | XP_011519632.1 | 11 | 1107 | |||
XM_011521331.2 | XP_011519633.1 | 10 | 989 | |||
XM_011521332.2 | XP_011519634.1 | 12 | 1230 | |||
XM_011521333.2 | XP_011519635.1 | 12 | 1247 | |||
XM_017021997.1 | XP_016877486.1 | 11 | 1190 | |||
XM_017021998.1 | XP_016877487.1 | 11 | 1414 | |||
GenBank mRNA | AK095072.1 | - | 11 | 1036 | ||
AK096501.1 | - | 2 | 565 | |||
AK303724.1 | - | 3 | 454 | |||
BC038451.1 | - | 11 | 1020 |