Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012744.1 | NP_001012762.1 | 2 | 778 | ||
XM_006244607.2 | XP_006244669.1 | 2 | 802 | |||
GenBank mRNA | BC090331.1 | - | 2 | 778 | 7300780 |