Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012968.2 | NP_001012986.2 | 1 | 2710 | ||
GenBank mRNA | AK021705.1 | - | 1 | 509 | ||
AK126931.1 | - | 1 | 2687 | |||
AL833314.1 | - | 1 | 2703 | |||
BC035088.1 | - | 1 | 923 | |||
BC043412.1 | - | 1 | 2222 | 6156224 |