Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013038.1 | NP_001013056.1 | 2 | 1375 | ||
GenBank mRNA | AB087405.1 | - | 1 | 155 | ||
AJ537426.2 | - | 2 | 1375 |