Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013094.1 | NP_001013112.1 | 6 | 843 | ||
GenBank mRNA | BC088316.1 | - | 6 | 843 | 7325177 | |
FQ211043.1 | - | 6 | 904 |