Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013130.1 | NP_001013148.1 | 11, 12 | 1474 | ||
XM_008770458.1 | XP_008768680.1 | 11, 12 | 3011 | |||
XM_008770460.1 | XP_008768682.1 | 10, 11 | 2868 | |||
XM_008770461.1 | XP_008768683.1 | 11, 12 | 1491 | |||
XM_008770462.1 | XP_008768684.1 | 10, 11 | 2868 | |||
XM_008770463.1 | XP_008768685.1 | 11, 12 | 3011 | |||
GenBank mRNA | AY238344.1 | - | 11, 12 | 1474 |