Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013130.1 | NP_001013148.1 | 24 | 4951 | ||
XM_008770458.1 | XP_008768680.1 | 23 | 6503 | |||
XM_008770460.1 | XP_008768682.1 | 22 | 6360 | |||
XM_008770461.1 | XP_008768683.1 | 23 | 4983 | |||
XM_008770462.1 | XP_008768684.1 | 22 | 6360 | |||
XM_008770463.1 | XP_008768685.1 | 23 | 6503 | |||
GenBank mRNA | AY238344.1 | - | 24 | 4951 |