Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013131.1 | NP_001013149.1 | 4 | 504 | ||
GenBank mRNA | BC091207.1 | - | 4 | 504 | 7381272 |