Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013162.1 | NP_001013180.1 | 6 | 850 | ||
XM_006234358.2 | XP_006234420.1 | 6 | 887 | |||
XM_006234359.2 | XP_006234421.1 | 6 | 889 | |||
XM_006234360.2 | XP_006234422.1 | 6 | 889 | |||
GenBank mRNA | BC088234.1 | - | 6 | 850 | 7324882 |