Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013164.3 | NP_001013182.3 | 3 | 293 | ||
XM_006233810.2 | XP_006233872.1 | 3 | 670 | |||
XM_006233811.2 | XP_006233873.1 | 3 | 763 | |||
XM_006233812.2 | XP_006233874.1 | 3 | 221 | |||
XM_006233813.2 | XP_006233875.2 | 3 | 233 | |||
XM_008761658.1 | XP_008759880.1 | 4 | 976 | |||
XM_008761659.1 | XP_008759881.1 | 4 | 980 | |||
GenBank mRNA | S61960.1 | - | 3 | 434 | ||
Z49696.1 | - | 4 | 442 |