Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEZF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018008.3 | NP_060478.3 | 2 | 480 | ||
GenBank mRNA | AF064845.1 | - | 2 | 318 | ||
AF332890.1 | - | 1 | 332 | |||
AK001004.1 | - | 1 | 392 | |||
AK290464.1 | - | 2 | 480 | |||
BC022464.1 | - | 2 | 311 | 4794121 | ||
BM930804.1 | - | 1 | 66 |