Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013184.1 | NP_001013202.1 | 6 | 642 | ||
GenBank mRNA | BC086423.1 | - | 6 | 642 | 7302011 | |
FQ220876.1 | - | 6 | 680 | |||
FQ222121.1 | - | 6 | 681 | |||
FQ230156.1 | - | 6 | 645 |
Gene Symbol: LOC100911295 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751029.3 | XP_003751077.1 | 6 | 713 |