Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013365.2 | NP_001013383.1 | 3 | 1051 | ||
GenBank mRNA | AK087945.1 | - | 1 | 461 | ||
AK155637.1 | - | 3 | 1002 |