Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lman2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013374.1 | NP_001013392.1 | 5 | 643 | ||
GenBank mRNA | AK041656.1 | - | 3 | 2488 | ||
AK142024.1 | - | 612 | ||||
AK158015.1 | - | 5 | 662 | |||
BC027226.1 | - | 4 | 561 | 2654285 | ||
BC046969.1 | - | 5 | 643 | 6313679 |