Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013742.3 | NP_001013764.1 | 3 | 831 | ||
XM_017029268.1 | XP_016884757.1 | 3 | 943 | |||
GenBank mRNA | AB183864.1 | - | 3 | 831 | ||
BC137319.1 | - | 3 | 815 | 9021316 | ||
BC137320.1 | - | 3 | 815 | 9021317 |