Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110316.2 | NP_001103786.2 | 2 | 494 | ||
GenBank mRNA | AK141250.1 | - | 2 | 496 | ||
CN715580.1 | - | 1 | 272 |
Gene Symbol: Zscan4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185173.1 | NP_001172102.1 | 2 | 494 |
Gene Symbol: Zscan4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013765.2 | NP_001013787.1 | 2 | 519 | ||
GenBank mRNA | BC050218.1 | - | 2 | 519 | 30009434 |
Gene Symbol: Zscan4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100186.1 | NP_001093656.1 | 2 | 508 | ||
GenBank mRNA | EF143406.1 | - | 2 | 508 |