Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diexf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013986.2 | NP_001014008.1 | 12 | 2269 | ||
XM_006250482.2 | XP_006250544.1 | 13 | 2459 | |||
GenBank mRNA | BC087114.1 | - | 12 | 2223 | 7193786 | |
FQ217506.1 | - | 13 | 2442 |