Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srd5a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013990.1 | NP_001014012.1 | 6, 7 | 803 | ||
GenBank mRNA | BC086584.1 | - | 6, 7 | 803 | 7307843 |