Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn2aip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014000.1 | NP_001014022.1 | 3 | 1379 | ||
XM_006253147.2 | XP_006253209.1 | 3 | 1248 | |||
GenBank mRNA | BC085832.1 | - | 3 | 1379 | 7134740 | |
BC098694.1 | - | 3 | 1142 |