Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mak16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014002.1 | NP_001014024.1 | 6, 7 | 500 | ||
GenBank mRNA | BC090035.1 | - | 6, 7 | 500 | 7367321 |